Learn More
MSC PRO-SAFE Lens, 4-1/4 x 2 in, Shade 11, 1 ea
Catalog No. 5000794648
Encompass
This fixed shade welding lens is designed to protect eyes during welding operations, providing a consistent shade level for optical clarity and safety. It meets stringent safety standards for reliable performance.
  • Fixed shade lens type
  • Window width: 4-1/4 in
  • Window height: 2 in
  • Lens shade: 11
  • Complies with ANSI Z87.1 standards
Catalog No. 50-007-94648 Supplier MSC Supplier No. 62674411
This fixed shade welding lens is designed to protect eyes during welding operations, providing a consistent shade level for optical clarity and safety. It meets stringent safety standards for reliable performance.
  • Fixed shade lens type
  • Window width: 4-1/4 in
  • Window height: 2 in
  • Lens shade: 11
  • Complies with ANSI Z87.1 standards