Learn More
MSC PRO-SAFE Magnifier, 1.5x Magnification, Optical Glass, Rectangular, Headband
Catalog No. 5000533286
Encompass
This headband magnifier is designed for applications requiring precise magnification. It features a 1.5x magnification with an optical glass lens, providing clear and consistent viewing. The rectangular lens shape and comfortable headband mounting make it suitable for prolonged use in various tasks.
  • Mounts securely with a headband
  • Provides 1.5x magnification
  • Features an optical glass lens
  • Has a rectangular lens shape
  • Offers a 20 inch focal distance
  • Does not include a light source
Catalog No. 50-005-33286 Supplier MSC Supplier No. 00322651
This headband magnifier is designed for applications requiring precise magnification. It features a 1.5x magnification with an optical glass lens, providing clear and consistent viewing. The rectangular lens shape and comfortable headband mounting make it suitable for prolonged use in various tasks.
  • Mounts securely with a headband
  • Provides 1.5x magnification
  • Features an optical glass lens
  • Has a rectangular lens shape
  • Offers a 20 inch focal distance
  • Does not include a light source