Learn More
MSC PRO-SAFE Mat, 10 x 3 ft., CFR Rubber, 1 ea
Catalog No. 5000802080
Encompass
This PRO-SAFE anti-fatigue mat is engineered for industrial environments, providing comfort and safety in areas with heavy-duty traffic. Its durable construction is suitable for both dry and wet conditions.
  • Beveled edges reduce trip hazards
  • Raised circle surface offers enhanced traction and comfort
  • Integrated drainage holes allow liquids to flow through
  • Constructed from CFR rubber for durability
  • Made with recycled content for environmental sustainability
Catalog No. 50-008-02080 Supplier MSC Supplier No. 88267232
This PRO-SAFE anti-fatigue mat is engineered for industrial environments, providing comfort and safety in areas with heavy-duty traffic. Its durable construction is suitable for both dry and wet conditions.
  • Beveled edges reduce trip hazards
  • Raised circle surface offers enhanced traction and comfort
  • Integrated drainage holes allow liquids to flow through
  • Constructed from CFR rubber for durability
  • Made with recycled content for environmental sustainability