Learn More
MSC PRO-SAFE Nomex Hood, Universal Size, 10 Cal/Square cm, 1 ea
Catalog No. 5000783046
Encompass
This flame-resistant Nomex hood is designed for arc flash protection. It offers a Hazard Risk Category of 2 and a maximum arc flash protection of 10 Cal/Square cm, making it suitable for environments requiring robust personal protective equipment.
  • Universal size for broad compatibility
  • Flame resistant Nomex material
  • Hazard Risk Category (HRC) 2 rating
  • Maximum arc flash protection of 10 Cal/Square cm
  • White color
Catalog No. 50-007-83046 Supplier MSC Supplier No. 77949873
This flame-resistant Nomex hood is designed for arc flash protection. It offers a Hazard Risk Category of 2 and a maximum arc flash protection of 10 Cal/Square cm, making it suitable for environments requiring robust personal protective equipment.
  • Universal size for broad compatibility
  • Flame resistant Nomex material
  • Hazard Risk Category (HRC) 2 rating
  • Maximum arc flash protection of 10 Cal/Square cm
  • White color