Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PRO-SAFE Plastic Chain, Yellow, 50 ft, 2 in, 1 ea

Catalog No. 5000792546
Encompass

This durable plastic chain is designed for effective crowd control and safety demarcation in various environments. Its bright yellow color ensures high visibility, making it ideal for directing pedestrian traffic and sectioning off hazardous or restricted areas.

  • Constructed from robust plastic for long-lasting performance
  • Features a highly visible yellow finish for safety applications
  • Measures 50 ft in length
  • Designed with a 2 in wide link
  • Type: Chain snap end

Catalog No. 50-007-92546 Supplier MSC Supplier No. 58393547
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This durable plastic chain is designed for effective crowd control and safety demarcation in various environments. Its bright yellow color ensures high visibility, making it ideal for directing pedestrian traffic and sectioning off hazardous or restricted areas.

  • Constructed from robust plastic for long-lasting performance
  • Features a highly visible yellow finish for safety applications
  • Measures 50 ft in length
  • Designed with a 2 in wide link
  • Type: Chain snap end

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.