Learn More
MSC PRO-SAFE Safety Glasses, Scratch-Resistant, Polycarbonate, Gray Lenses, 1 ea
Catalog No. 5000685219
Encompass
PRO-SAFE Safety Glasses feature scratch-resistant, polycarbonate, gray lenses, and a frameless design. They have non-adjustable temples and meet ANSI Z87.1+ standards.
  • Scratch-resistant lens coating
  • Polycarbonate lens material
  • Gray lenses
  • Frameless style with wrap around design
  • Non-adjustable temples
  • UV protection
  • Flexible temple type
  • Universal size
Catalog No. 50-006-85219 Supplier MSC Supplier No. 76564657
PRO-SAFE Safety Glasses feature scratch-resistant, polycarbonate, gray lenses, and a frameless design. They have non-adjustable temples and meet ANSI Z87.1+ standards.
  • Scratch-resistant lens coating
  • Polycarbonate lens material
  • Gray lenses
  • Frameless style with wrap around design
  • Non-adjustable temples
  • UV protection
  • Flexible temple type
  • Universal size