Learn More
MSC PRO-SAFE Safety Glasses, Scratch-Resistant, Polycarbonate, Silver Lenses, 1 ea
Catalog No. 5000688373
Encompass
These safety glasses feature a silver frame, non-adjustable temples, and are part of the Mac 3 series. They meet ANSI Z87.1+ standards for eye protection.
  • Tinted lenses
  • Scratch-resistant lens coating
  • Frameless design
  • Wrap-around frame for comprehensive protection
  • Silver polycarbonate lenses and frame
  • Universal size
  • Clear, flexible temples
  • UV protection
  • Meets ANSI Z87.1+ standards
Catalog No. 50-006-88373 Supplier MSC Supplier No. 93884369
These safety glasses feature a silver frame, non-adjustable temples, and are part of the Mac 3 series. They meet ANSI Z87.1+ standards for eye protection.
  • Tinted lenses
  • Scratch-resistant lens coating
  • Frameless design
  • Wrap-around frame for comprehensive protection
  • Silver polycarbonate lenses and frame
  • Universal size
  • Clear, flexible temples
  • UV protection
  • Meets ANSI Z87.1+ standards