Learn More
MSC PRO-SAFE Safety Goggles, Chemical Splash, Indirect Vent, 1 ea
Catalog No. 5000675622
Encompass
PRO-SAFE safety goggles provide reliable protection against chemical splashes. These goggles feature a clear polycarbonate lens and indirect ventilation for enhanced safety and comfort.
  • Uncoated clear polycarbonate lens
  • Indirect ventilation protects against chemical splash
  • Universal size fits comfortably over glasses
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes a durable strap
  • Constructed with a clear vinyl frame
Catalog No. 50-006-75622 Supplier MSC Supplier No. 89374482
PRO-SAFE safety goggles provide reliable protection against chemical splashes. These goggles feature a clear polycarbonate lens and indirect ventilation for enhanced safety and comfort.
  • Uncoated clear polycarbonate lens
  • Indirect ventilation protects against chemical splash
  • Universal size fits comfortably over glasses
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes a durable strap
  • Constructed with a clear vinyl frame