Learn More
MSC PRO-SAFE Safety Goggles, Chemical Splash, Uncoated, Polycarbonate Lenses, 1 ea
Catalog No. 5000769071
Encompass
These safety goggles are designed to provide protection against chemical splash. They feature clear, uncoated polycarbonate lenses and a direct vent system. The goggles have a green PVC frame, are universal in size, and offer UV protection. They are not intended for use over prescription glasses.
  • Clear lens
  • Uncoated lens
  • Direct vent system
  • Chemical protection
  • Polycarbonate lens material
  • Green frame
  • PVC frame material
  • Universal size
  • Not designed for use over prescription glasses
  • UV protection
Catalog No. 50-007-69071 Supplier MSC Supplier No. 56602022
These safety goggles are designed to provide protection against chemical splash. They feature clear, uncoated polycarbonate lenses and a direct vent system. The goggles have a green PVC frame, are universal in size, and offer UV protection. They are not intended for use over prescription glasses.
  • Clear lens
  • Uncoated lens
  • Direct vent system
  • Chemical protection
  • Polycarbonate lens material
  • Green frame
  • PVC frame material
  • Universal size
  • Not designed for use over prescription glasses
  • UV protection