Learn More
MSC PRO-SAFE Safety Goggles, Dust, Scratch-Resistant, Polycarbonate, Direct Vent, 1 ea
Catalog No. 5000789616
Encompass
These safety goggles offer protection against dust with clear, scratch-resistant polycarbonate lenses and a direct vent system. Featuring a clear frame, they are universal in size and fit over prescription glasses, also providing UV protection.
  • Scratch-resistant lens coating
  • Direct ventilation
  • Dust protection
  • Clear polycarbonate lenses
  • Clear vinyl and plastic frame
  • Universal size fits over glasses
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes strap
Catalog No. 50-007-89616 Supplier MSC Supplier No. 89374474
These safety goggles offer protection against dust with clear, scratch-resistant polycarbonate lenses and a direct vent system. Featuring a clear frame, they are universal in size and fit over prescription glasses, also providing UV protection.
  • Scratch-resistant lens coating
  • Direct ventilation
  • Dust protection
  • Clear polycarbonate lenses
  • Clear vinyl and plastic frame
  • Universal size fits over glasses
  • Provides UV protection
  • Meets ANSI Z87.1+ standards
  • Includes strap