Learn More
MSC PRO-SAFE Sorbent Roll, Universal, Extra Heavy, 150 ft
Catalog No. 5000794903
Encompass
This Sorbent Roll is designed for universal use, featuring an extra heavy weight and a capacity of 56 gal.. It is yellow in color and perforated.
  • Dual use: functions as both a universal sorbent and a caution slip-hazard warning.
  • Hydrophilic: absorbs all liquids, including oil, water, coolants, and chemicals.
  • Common indoor use: suitable for cleaning up or controlling any spill, leak, or drip indoors.
  • Material: made from polypropylene.
  • Absorbency weight: extra heavy.
  • Perforated: yes.
Catalog No. 50-007-94903 Supplier MSC Supplier No. 42099960
This Sorbent Roll is designed for universal use, featuring an extra heavy weight and a capacity of 56 gal.. It is yellow in color and perforated.
  • Dual use: functions as both a universal sorbent and a caution slip-hazard warning.
  • Hydrophilic: absorbs all liquids, including oil, water, coolants, and chemicals.
  • Common indoor use: suitable for cleaning up or controlling any spill, leak, or drip indoors.
  • Material: made from polypropylene.
  • Absorbency weight: extra heavy.
  • Perforated: yes.