Learn More
MSC PRO-SAFE Sorbent Roll, Universal Use, Polypropylene, 150 ft, 1 ea
Catalog No. 5000782404
Encompass
This sorbent roll is designed for universal use, effectively absorbing a wide range of industrial liquids. Constructed from durable polypropylene, it provides a reliable solution for managing spills and maintaining a safe work environment.
  • Suitable for absorbing oils, coolants, solvents, and water-based liquids
  • Made from high-quality polypropylene for superior absorption
  • Large capacity for containing significant spills
  • Versatile for use as a walk-on mat or for covering extensive areas
Catalog No. 50-007-82404 Supplier MSC Supplier No. 04019931
This sorbent roll is designed for universal use, effectively absorbing a wide range of industrial liquids. Constructed from durable polypropylene, it provides a reliable solution for managing spills and maintaining a safe work environment.
  • Suitable for absorbing oils, coolants, solvents, and water-based liquids
  • Made from high-quality polypropylene for superior absorption
  • Large capacity for containing significant spills
  • Versatile for use as a walk-on mat or for covering extensive areas