Learn More
MSC PRO-SAFE Tacky Clean Room Mat, 36 x 60 in., 4/Pack
Catalog No. 5000673330
Encompass
The Clean Room Mat features tacky sheets designed for use in dry environments. It measures 36 in. wide and 60 in. long, containing 30 layers per mat. These mats are white with a smooth surface pattern and are not reusable.
  • Tacky layers: 30 per mat
  • Designed for dry environments
  • Smooth surface pattern
  • Not reusable
  • White color
  • Made in China
  • Supplied as 4 mats per pack
Catalog No. 50-006-73330 Supplier MSC Supplier No. 53136883
The Clean Room Mat features tacky sheets designed for use in dry environments. It measures 36 in. wide and 60 in. long, containing 30 layers per mat. These mats are white with a smooth surface pattern and are not reusable.
  • Tacky layers: 30 per mat
  • Designed for dry environments
  • Smooth surface pattern
  • Not reusable
  • White color
  • Made in China
  • Supplied as 4 mats per pack