Learn More
MSC PRO-SAFE Terry Heat Resistant Glove, Size L, 1 Pair
Catalog No. 5000765838
Encompass
This terry heat resistant glove is designed for applications requiring protection against heat. It features a knit wrist for a secure fit and is suitable for various industrial tasks.
  • Heat resistant glove type
  • Unlined construction
  • Overall length of 10.6 in.
  • Natural color, supplied as a pair
Catalog No. 50-007-65838 Supplier MSC Supplier No. 93880276
This terry heat resistant glove is designed for applications requiring protection against heat. It features a knit wrist for a secure fit and is suitable for various industrial tasks.
  • Heat resistant glove type
  • Unlined construction
  • Overall length of 10.6 in.
  • Natural color, supplied as a pair