Learn More
MSC PRO-SAFE Terry Heat Resistant Glove, Size S, 1 Pair
Catalog No. 5000766109
Encompass
This terry heat resistant glove is designed for general purpose hand protection, offering thermal resistance for various applications. It features a knit wrist for a secure fit and is supplied as a pair.
  • Heat resistant
  • Size S
  • Knit wrist
  • Gray color
  • Unlined for dexterity
  • Made in UNITED STATES
Catalog No. 50-007-66109 Supplier MSC Supplier No. 93880300
This terry heat resistant glove is designed for general purpose hand protection, offering thermal resistance for various applications. It features a knit wrist for a secure fit and is supplied as a pair.
  • Heat resistant
  • Size S
  • Knit wrist
  • Gray color
  • Unlined for dexterity
  • Made in UNITED STATES