Learn More
MSC PRO-SAFE Unlined Drivers Gloves, Top Grain Cowhide, Small, 1 Pair
Catalog No. 5000796373
Encompass
These unlined drivers gloves are crafted from premium grade top grain cowhide, offering a smooth leather palm for durability and dexterity, complemented by a split cowhide back for comfort, breathability, and excellent abrasion resistance.
  • Top grain cowhide leather palm provides durability and dexterity.
  • Split cowhide leather back ensures comfort, durability, and abrasion resistance.
  • Shoulder split cowhide back for economical construction.
  • Keystone thumb reduces hand fatigue and enhances dexterity.
  • Stitched with para-aramid thread for strength and burn resistance.
  • Slip-on styling for a good fit and comfort.
  • Elastic closure for safety and comfort.
Catalog No. 50-007-96373 Supplier MSC Supplier No. 06016893
These unlined drivers gloves are crafted from premium grade top grain cowhide, offering a smooth leather palm for durability and dexterity, complemented by a split cowhide back for comfort, breathability, and excellent abrasion resistance.
  • Top grain cowhide leather palm provides durability and dexterity.
  • Split cowhide leather back ensures comfort, durability, and abrasion resistance.
  • Shoulder split cowhide back for economical construction.
  • Keystone thumb reduces hand fatigue and enhances dexterity.
  • Stitched with para-aramid thread for strength and burn resistance.
  • Slip-on styling for a good fit and comfort.
  • Elastic closure for safety and comfort.