Learn More
MSC PRO-SAFE Work Gloves, Latex Crinkle Grip, Medium, 1 Pair
Catalog No. 5000793328
Encompass
These work gloves are designed for general purpose applications, featuring a cotton blend material and a latex coating on the palm and fingers for a crinkle grip.
  • General purpose application
  • Cotton blend material
  • Unlined with knit cuff
  • Latex coating on palm and fingers
  • Crinkle grip surface
  • Medium size (numeric size 8)
  • Overall length 10 in
  • Seamless construction
Catalog No. 50-007-93328 Supplier MSC Supplier No. 84492156
These work gloves are designed for general purpose applications, featuring a cotton blend material and a latex coating on the palm and fingers for a crinkle grip.
  • General purpose application
  • Cotton blend material
  • Unlined with knit cuff
  • Latex coating on palm and fingers
  • Crinkle grip surface
  • Medium size (numeric size 8)
  • Overall length 10 in
  • Seamless construction