Learn More
MSC PRO-SAFE Work Gloves, PVC-Coated, Polyester, Small, General Purpose, 1 Pair
Catalog No. 5000793879
Encompass
PRO-SAFE work gloves are designed for general purpose applications. These gloves feature a polyester material with PVC coating, providing durability and protection for various tasks.
  • General purpose glove type
  • Made from polyester material
  • Unlined for comfort
  • Knit cuff material
  • PVC coating for enhanced grip and protection
  • Full coating coverage
  • Small glove size (numeric size 7)
  • Orange color for visibility
  • Overall length of 9.7 in
  • Seamless construction
Catalog No. 50-007-93879 Supplier MSC Supplier No. 70347257
PRO-SAFE work gloves are designed for general purpose applications. These gloves feature a polyester material with PVC coating, providing durability and protection for various tasks.
  • General purpose glove type
  • Made from polyester material
  • Unlined for comfort
  • Knit cuff material
  • PVC coating for enhanced grip and protection
  • Full coating coverage
  • Small glove size (numeric size 7)
  • Orange color for visibility
  • Overall length of 9.7 in
  • Seamless construction