Learn More
MSC PRO-SAFE Work Gloves, Size Medium, Polyurethane-Coated, Nylon, 1 Pair
Catalog No. 5000675593
Encompass
These work gloves are designed for general purpose applications. They are size medium, black in color, and measure 9.3 in. long. The gloves are made from nylon with a knit cuff and feature a polyurethane coating on the palm and fingers. They are unlined and seamless, and come as a pair.
  • General purpose glove type and application
  • Nylon glove material with knit cuff
  • Polyurethane coating on palm and fingers
  • Medium size (numeric size 8)
  • Black color
  • Overall length 9.3 in.
  • Unlined and seamless design
  • Sold as a pair
  • Made in United States
Catalog No. 50-006-75593 Supplier MSC Supplier No. 84492073
These work gloves are designed for general purpose applications. They are size medium, black in color, and measure 9.3 in. long. The gloves are made from nylon with a knit cuff and feature a polyurethane coating on the palm and fingers. They are unlined and seamless, and come as a pair.
  • General purpose glove type and application
  • Nylon glove material with knit cuff
  • Polyurethane coating on palm and fingers
  • Medium size (numeric size 8)
  • Black color
  • Overall length 9.3 in.
  • Unlined and seamless design
  • Sold as a pair
  • Made in United States