Learn More
MSC PRO-SAFE Disposable Coveralls, Chemical Resistant, Film Laminate, 4X-Large, 25/Pk.
Catalog No. 5000685822
Encompass
These chemical-resistant disposable coveralls are designed for reliable protection in environments where exposure to hazardous materials is a concern. Constructed from a durable film laminate, the garment offers a robust barrier while maintaining comfort for the wearer. The design features a convenient zipper closure and elastic cuffs and ankles to ensure a secure fit and minimize the entry of contaminants.
  • Chemical-resistant film laminate material
  • Size 4X-Large
  • Material weight of 1.87 oz
  • Elastic cuffs and ankles for secure coverage
  • Zipper closure for easy donning and removal
  • Serged seams
  • ANSI/ISEA 101-1996 compliant
  • ISO class 5 rated
Catalog No. 50-006-85822 Supplier MSC Supplier No. 61048211
These chemical-resistant disposable coveralls are designed for reliable protection in environments where exposure to hazardous materials is a concern. Constructed from a durable film laminate, the garment offers a robust barrier while maintaining comfort for the wearer. The design features a convenient zipper closure and elastic cuffs and ankles to ensure a secure fit and minimize the entry of contaminants.
  • Chemical-resistant film laminate material
  • Size 4X-Large
  • Material weight of 1.87 oz
  • Elastic cuffs and ankles for secure coverage
  • Zipper closure for easy donning and removal
  • Serged seams
  • ANSI/ISEA 101-1996 compliant
  • ISO class 5 rated