Learn More
MSC Instant Power Professional Septic System Treatment, Unscented, 5 gal
Catalog No. 502819647
Encompass
This environmentally safe Septic System Treatment formula is highly effective in keeping septic systems running efficiently and trouble-free. Its microbial blend helps digest system-clogging matter, maintains lateral/leach field lines, and restores porosity to drain fields.
  • Dissolves and liquefies waste
  • Maintains ideal bacterial balance
  • Reduces costly pump outs
  • Controls odors
  • Biodegradable and unscented
  • Safe for all pipes and septic systems
  • Compatible with plastic and metal pipe material
  • Septic system and garbage disposal safe
  • Ready to use formula
Catalog No. 50-281-9647 Supplier MSC Supplier No. 14965503
This environmentally safe Septic System Treatment formula is highly effective in keeping septic systems running efficiently and trouble-free. Its microbial blend helps digest system-clogging matter, maintains lateral/leach field lines, and restores porosity to drain fields.
  • Dissolves and liquefies waste
  • Maintains ideal bacterial balance
  • Reduces costly pump outs
  • Controls odors
  • Biodegradable and unscented
  • Safe for all pipes and septic systems
  • Compatible with plastic and metal pipe material
  • Septic system and garbage disposal safe
  • Ready to use formula