Learn More
MSC PRO-SOURCE Ball Bearing Trolley, 1 Ton Capacity, 3-5/32 to 5-3/4 in Flange
Catalog No. 5000528696
Encompass
This ball bearing trolley is designed for efficient material handling, offering a 1-ton load capacity and a wide range design to accommodate various I-beam sizes.
  • Offers a load capacity of 1 ton (2000 lbs).
  • Accommodates flange widths from 3-5/32 to 5-3/4 in.
  • Designed for I-beams with a height range of 5 to 24 in.
  • Features a wheel diameter of 2-5/8 in.
  • Minimum radius of 35 in.
  • Wide range ball bearing type.
Catalog No. 50-005-28696 Supplier MSC Supplier No. 79487559
This ball bearing trolley is designed for efficient material handling, offering a 1-ton load capacity and a wide range design to accommodate various I-beam sizes.
  • Offers a load capacity of 1 ton (2000 lbs).
  • Accommodates flange widths from 3-5/32 to 5-3/4 in.
  • Designed for I-beams with a height range of 5 to 24 in.
  • Features a wheel diameter of 2-5/8 in.
  • Minimum radius of 35 in.
  • Wide range ball bearing type.