Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC PRO-SOURCE Contractor Bags, 45 gal, 1.5 mil, Flat Pack, 100/Pk.

Catalog No. 502867893
Encompass

PRO-SOURCE contractor trash bags are designed for heavy-duty waste containment. Made from durable polyethylene, these bags are suitable for various professional and industrial applications, including construction sites and general clean-up.

  • Capacity: 45 gal
  • Overall thickness: 1.5 mil
  • Material: Polyethylene
  • Contains recycled material
  • Color: Black
  • Closure type: Tie
  • Dispensing type: Flat pack
  • Unscented
  • Bottom type: Flat
  • SCS certified for sustainability

Catalog No. 50-286-7893 Supplier MSC Supplier No. 42107904
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PRO-SOURCE contractor trash bags are designed for heavy-duty waste containment. Made from durable polyethylene, these bags are suitable for various professional and industrial applications, including construction sites and general clean-up.

  • Capacity: 45 gal
  • Overall thickness: 1.5 mil
  • Material: Polyethylene
  • Contains recycled material
  • Color: Black
  • Closure type: Tie
  • Dispensing type: Flat pack
  • Unscented
  • Bottom type: Flat
  • SCS certified for sustainability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.