Learn More
MSC PRO-SOURCE Bottle with Applicator, 12 oz, HDPE, 38/400, 1 ea
Catalog No. 502866882
Encompass
This clear, 12 oz HDPE cylinder bottle comes with an applicator tip, suitable for various applications including cleaning, hair care, and food service products.
  • Includes applicator tip
  • Cylinder bottle design
  • Container capacity: 12 oz
  • Clear color
  • Screw closure size: 38/400
  • For use with cleaning products, hair care products, and food service products
Catalog No. 50-286-6882 Supplier MSC Supplier No. 30979009
This clear, 12 oz HDPE cylinder bottle comes with an applicator tip, suitable for various applications including cleaning, hair care, and food service products.
  • Includes applicator tip
  • Cylinder bottle design
  • Container capacity: 12 oz
  • Clear color
  • Screw closure size: 38/400
  • For use with cleaning products, hair care products, and food service products