Learn More
MSC PRO-SOURCE Drum Caddy, 55 gal, 1 Drum, 1,250 lb Capacity
Catalog No. 5000528226
Encompass
This drum caddy is designed for transporting and storing a single 55 gallon drum. It features a robust metal construction and can safely handle loads up to 1,250 lb. Its durable design ensures reliable performance for various industrial applications.
  • Designed for one 55 gallon drum
  • 1,250 lb load capacity
  • Constructed from metal for durability
  • 23.5 in outside diameter
Catalog No. 50-005-28226 Supplier MSC Supplier No. 48303630
This drum caddy is designed for transporting and storing a single 55 gallon drum. It features a robust metal construction and can safely handle loads up to 1,250 lb. Its durable design ensures reliable performance for various industrial applications.
  • Designed for one 55 gallon drum
  • 1,250 lb load capacity
  • Constructed from metal for durability
  • 23.5 in outside diameter