Learn More
MSC PRO-SOURCE Drum Caddy, 55 gal Drum, 1,400 Lb Capacity, Steel
Catalog No. 5000527069
Encompass
This caddy is designed to facilitate the safe and efficient transport of drums within industrial and laboratory settings. It accommodates a single 55-gallon drum and has a substantial load capacity, constructed from durable steel for long-lasting performance.
  • Accommodates one 55 gallon drum.
  • Provides a load capacity of 1,400 lb.
  • Constructed from steel for durability.
Catalog No. 50-005-27069 Supplier MSC Supplier No. 48303622
This caddy is designed to facilitate the safe and efficient transport of drums within industrial and laboratory settings. It accommodates a single 55-gallon drum and has a substantial load capacity, constructed from durable steel for long-lasting performance.
  • Accommodates one 55 gallon drum.
  • Provides a load capacity of 1,400 lb.
  • Constructed from steel for durability.