Learn More
MSC PRO-SOURCE Drum Grab, 1 Drum, 800 lb Capacity, Fork Mounted
Catalog No. 5000529201
Encompass
The PRO-SOURCE Drum Grab is a fork-mounted tool designed for handling drums.
  • Designed to handle one drum.
  • Fork mounted drum grab.
  • Has a load capacity of 800 lb.
  • Suitable for drums with a capacity of 55 gallons.
  • Country of origin is China.
Catalog No. 50-005-29201 Supplier MSC Supplier No. 20243010
The PRO-SOURCE Drum Grab is a fork-mounted tool designed for handling drums.
  • Designed to handle one drum.
  • Fork mounted drum grab.
  • Has a load capacity of 800 lb.
  • Suitable for drums with a capacity of 55 gallons.
  • Country of origin is China.