Learn More
MSC PRO-SOURCE Industrial Circulation Fan, 20 in Dia, 1/4 hp, 2 Speed
Catalog No. 5000509126
Encompass
This industrial circulation fan is designed for efficient air movement in various settings. Featuring a 20 in diameter and a 1/4 hp motor, it offers two-speed operation and single-phase power. Its I-beam mounting design ensures secure installation.
  • 20 in fan diameter
  • 1/4 hp motor
  • Single phase operation
  • Two speed settings
  • Designed for I-beam mounting
Catalog No. 50-005-09126 Supplier MSC Supplier No. 94463676
This industrial circulation fan is designed for efficient air movement in various settings. Featuring a 20 in diameter and a 1/4 hp motor, it offers two-speed operation and single-phase power. Its I-beam mounting design ensures secure installation.
  • 20 in fan diameter
  • 1/4 hp motor
  • Single phase operation
  • Two speed settings
  • Designed for I-beam mounting