Learn More
MSC PRO-SOURCE Industrial Powered Pallet Truck, 5500 lb, 27 x 48 in
Catalog No. 5000529304
Encompass
This industrial powered pallet truck offers a robust 5500 lb load capacity, designed for efficient material handling. It features precise control with foot operation and is suitable for various warehouse and industrial applications.
  • 5500 lb load capacity
  • 27 in overall width
  • 48 in fork length
  • 3-3/8 in lowered lifting height
  • 7-7/8 in maximum lift height
  • Foot control type
Catalog No. 50-005-29304 Supplier MSC Supplier No. 20242996
This industrial powered pallet truck offers a robust 5500 lb load capacity, designed for efficient material handling. It features precise control with foot operation and is suitable for various warehouse and industrial applications.
  • 5500 lb load capacity
  • 27 in overall width
  • 48 in fork length
  • 3-3/8 in lowered lifting height
  • 7-7/8 in maximum lift height
  • Foot control type