Learn More
MSC PRO-SOURCE Pedestal Fan, 24 in, 120V, Single Speed, 1 ea
Catalog No. 5000505877
Encompass
This PRO-SOURCE pedestal fan features a single speed setting and a 24-inch blade, delivering an airflow of 10480 CFM and operating at 2.8A.
  • Single speed setting
  • A 24-inch blade diameter
  • Airflow of 10480 CFM
  • Operates at 120V
  • Non-oscillating design
  • Aluminum blade material
  • White color
  • Direct drive type
  • Suitable for outdoor use
Catalog No. 50-005-05877 Supplier MSC Supplier No. 49698590
This PRO-SOURCE pedestal fan features a single speed setting and a 24-inch blade, delivering an airflow of 10480 CFM and operating at 2.8A.
  • Single speed setting
  • A 24-inch blade diameter
  • Airflow of 10480 CFM
  • Operates at 120V
  • Non-oscillating design
  • Aluminum blade material
  • White color
  • Direct drive type
  • Suitable for outdoor use