Learn More
MSC PRO-SOURCE Platform Truck, Aluminum, 36 x 24 in, 2000 lb Capacity
Catalog No. 5000536712
Encompass
This platform truck is designed with an aluminum construction for durability and a 2000 lb load capacity. It features 8-inch diameter casters for easy mobility.
  • Aluminum construction for durability.
  • 2000 lb load capacity.
  • 8-inch diameter casters for mobility.
Catalog No. 50-005-36712 Supplier MSC Supplier No. 89739080
This platform truck is designed with an aluminum construction for durability and a 2000 lb load capacity. It features 8-inch diameter casters for easy mobility.
  • Aluminum construction for durability.
  • 2000 lb load capacity.
  • 8-inch diameter casters for mobility.