Learn More
MSC PRO-SOURCE Portable Electric Salamander Heater, 30KW, 480VAC, 3PH
Catalog No. 5000507942
Encompass
The PRO-SOURCE Portable Electric Salamander Heater is a robust heating solution designed for various applications. It operates on 480V three-phase power, delivers 30 kW of heating, and provides 1900 CFM.
  • Built-in thermostat
  • Overheat protection
  • Rubber wheels for easy moving on any floor
Catalog No. 50-005-07942 Supplier MSC Supplier No. 98490683
The PRO-SOURCE Portable Electric Salamander Heater is a robust heating solution designed for various applications. It operates on 480V three-phase power, delivers 30 kW of heating, and provides 1900 CFM.
  • Built-in thermostat
  • Overheat protection
  • Rubber wheels for easy moving on any floor