Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Pro-Source™ Round Trash Can Lid

Catalog No. 501298213
Encompass
Change view
Click to view available options
Pro-Source™ Round Trash Can Lid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-129-8213 Pro-Source™ Round Trash Can Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-129-8213 Supplier MSC Supplier No. 66506536
This item is not returnable. View return policy

Specifications

Color Red
Shape Round
For Use With (Equipment) 32gal. trash cans
Width (English) 22.25 in.
Height (English) 1.375 in.
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.