Learn More
MSC PRO-SOURCE Shell Pulley, 1,400 lb Load Limit, Double Sheave, Wood
Catalog No. 5000513586
Encompass
This shell pulley is designed with a 1,400 lb load limit. It features a double sheave configuration and is constructed from wood, suitable for fibrous rope applications.
  • Double sheave design.
  • 1,400 lb load limit.
  • Compatible with fibrous rope.
  • Constructed from wood.
  • Features a 2-1/4 inch outside diameter.
  • Accommodates 1/2 inch diameter rope.
Catalog No. 50-005-13586 Supplier MSC Supplier No. 09431453
This shell pulley is designed with a 1,400 lb load limit. It features a double sheave configuration and is constructed from wood, suitable for fibrous rope applications.
  • Double sheave design.
  • 1,400 lb load limit.
  • Compatible with fibrous rope.
  • Constructed from wood.
  • Features a 2-1/4 inch outside diameter.
  • Accommodates 1/2 inch diameter rope.