Learn More
MSC PRO-SOURCE Side Release Snatch Block, 17,600 lb, 6 in OD
Catalog No. 5000535249
Encompass
This side release snatch block has a load limit of 17,600 lb. It features a single sheave with a 6 in outside diameter and is designed for use with a 5/8 in wire rope. The snatch block is constructed from steel.
  • Load limit of 17,600 lb
  • Single sheave
  • 6 in outside diameter
  • Designed for 5/8 in wire rope
  • Constructed from steel
Catalog No. 50-005-35249 Supplier MSC Supplier No. 09431719
This side release snatch block has a load limit of 17,600 lb. It features a single sheave with a 6 in outside diameter and is designed for use with a 5/8 in wire rope. The snatch block is constructed from steel.
  • Load limit of 17,600 lb
  • Single sheave
  • 6 in outside diameter
  • Designed for 5/8 in wire rope
  • Constructed from steel