Learn More
MSC PRO-SOURCE Snatch Block, 8,800 Lbs. Load Limit, Side Release, 4-1/2 in Outside Diameter, Steel
Catalog No. 5000536239
Encompass
This snatch block features a single sheave and a side release mechanism, designed for heavy-duty applications. It has a load limit of 8,800 pounds and is constructed from steel, suitable for use with 3/8 inch diameter wire rope.
  • Single sheave design for efficient operation.
  • 8,800 Lbs. load limit for demanding tasks.
  • Side release mechanism for easy rope insertion and removal.
  • 4-1/2 in outside diameter.
  • Compatible with 3/8 in diameter wire rope.
  • Durable steel construction.
Catalog No. 50-005-36239 Supplier MSC Supplier No. 09431768
This snatch block features a single sheave and a side release mechanism, designed for heavy-duty applications. It has a load limit of 8,800 pounds and is constructed from steel, suitable for use with 3/8 inch diameter wire rope.
  • Single sheave design for efficient operation.
  • 8,800 Lbs. load limit for demanding tasks.
  • Side release mechanism for easy rope insertion and removal.
  • 4-1/2 in outside diameter.
  • Compatible with 3/8 in diameter wire rope.
  • Durable steel construction.