Learn More
MSC PRO-SOURCE Welded Chain, 5/16, Short Link, Grade 70, Yellow
Catalog No. 5000529219
Encompass
The PRO-SOURCE Welded Chain is a Grade 70, short link chain, featuring a 5/16 trade size and a yellow finish. This durable chain is designed for various demanding applications.
  • 5/16 trade size for general purpose use
  • Short link design for flexibility
  • Grade 70 for high strength and durability
  • Yellow finish for visibility and corrosion resistance
  • Overall length of 16 ft for extended reach
  • Inside width of 0.50 in
  • Inside length of 1.10 in
Catalog No. 50-005-29219 Supplier MSC Supplier No. 15821283
The PRO-SOURCE Welded Chain is a Grade 70, short link chain, featuring a 5/16 trade size and a yellow finish. This durable chain is designed for various demanding applications.
  • 5/16 trade size for general purpose use
  • Short link design for flexibility
  • Grade 70 for high strength and durability
  • Yellow finish for visibility and corrosion resistance
  • Overall length of 16 ft for extended reach
  • Inside width of 0.50 in
  • Inside length of 1.10 in