Learn More
MSC Diversey High Efficiency Floor Stripper, Green Seal Certified, 5 L
Catalog No. 502819326
Encompass
Pro Strip™ SC High Efficiency Floor Stripper is a highly concentrated, low odor floor stripper formulated to meet Green Seal® requirements. It is designed for floor finish removal on VCT, terrazzo, and concrete flooring, and can also be used on tile and grout.
  • Powerful, fast-acting, low foam formula
  • Quickly penetrates layers of finish upon application
  • Delivers fast, consistent removal of floor finish
  • Green Seal® Certification
  • Suitable for facilities where odor is a concern
  • Not formulated with butyl or ammonia
  • Water-based, concentrated solution
  • PH of 11.5 or greater
Catalog No. 50-281-9326 Supplier MSC Supplier No. 03849734
Pro Strip™ SC High Efficiency Floor Stripper is a highly concentrated, low odor floor stripper formulated to meet Green Seal® requirements. It is designed for floor finish removal on VCT, terrazzo, and concrete flooring, and can also be used on tile and grout.
  • Powerful, fast-acting, low foam formula
  • Quickly penetrates layers of finish upon application
  • Delivers fast, consistent removal of floor finish
  • Green Seal® Certification
  • Suitable for facilities where odor is a concern
  • Not formulated with butyl or ammonia
  • Water-based, concentrated solution
  • PH of 11.5 or greater