Learn More
Medchemexpress LLC PRO20 | 99.2% | 2341.82 | 25 MG
SDP
Catalog No. 5000425964
Encompass_Preferred
PRO20 is a specific and competitive (pro)renin receptor (PRR) antagonist. It inhibits Prorenin-induced calcium influx with an IC50 value of 81 nmol/L. PRO20 works by blocking the binding of prorenin to the PRR, which in turn inhibits the activation of the renin-angiotensin system (RAS) and reduces the production of angiotensin II (Ang II). It is considered promising for research into antihypertensive agents.
  • Specific and competitive (pro)renin receptor (PRR) antagonist
  • Inhibits prorenin-induced calcium influx
  • Reduces the production of angiotensin II (Ang II)
  • Promising for research into antihypertensive agents
Catalog No. 50-004-25964 Supplier Medchemexpress LLC Supplier No. 25MGHYP10933
PRO20 is a specific and competitive (pro)renin receptor (PRR) antagonist. It inhibits Prorenin-induced calcium influx with an IC50 value of 81 nmol/L. PRO20 works by blocking the binding of prorenin to the PRR, which in turn inhibits the activation of the renin-angiotensin system (RAS) and reduces the production of angiotensin II (Ang II). It is considered promising for research into antihypertensive agents.
  • Specific and competitive (pro)renin receptor (PRR) antagonist
  • Inhibits prorenin-induced calcium influx
  • Reduces the production of angiotensin II (Ang II)
  • Promising for research into antihypertensive agents