Learn More
Lonza Walkersville Pro293 a Serum-free Medium – Chemically Defined for 293 Adherent Cells
Catalog No. 502120215
Encompass
Pro293a Chemically Defined, Serum-free Media were optimized to support high-density growth and recombinant protein production in 293 neonatal kidney cells. They are chemically defined to ease regulatory compliance and downstream protein purification.They contain very low levels of recombinant human insulin, and are free of animal-origin components.Pro293s Medium — for 293 cells growing in suspension culturePro293 a Medium — for 293 cells growing in adherent cultureBoth media do not contain L-glutamine or phenol red. Pluronic F-68 is present at 0.1%.This BioWhittaker Cell Culture Media is produced according to cGMP and labelled "For Further Manufacturing" (FFM). If you require different pack sizes, contact Lonza for more details.Benefits:Serum FreeChemically DefinedLow protein contentNon Animal originApplications:Serum-free Cell CultureContent:1L bottle
Catalog No. 50-212-0215 Supplier Lonza Walkersville Supplier No. BEBP12764Q
Pro293a Chemically Defined, Serum-free Media were optimized to support high-density growth and recombinant protein production in 293 neonatal kidney cells. They are chemically defined to ease regulatory compliance and downstream protein purification.They contain very low levels of recombinant human insulin, and are free of animal-origin components.Pro293s Medium — for 293 cells growing in suspension culturePro293 a Medium — for 293 cells growing in adherent cultureBoth media do not contain L-glutamine or phenol red. Pluronic F-68 is present at 0.1%.This BioWhittaker Cell Culture Media is produced according to cGMP and labelled "For Further Manufacturing" (FFM). If you require different pack sizes, contact Lonza for more details.Benefits:Serum FreeChemically DefinedLow protein contentNon Animal originApplications:Serum-free Cell CultureContent:1L bottle