Learn More
CPC Scientific H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Pro-Arg-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP107B
Encompass
Selective NPY agonist at Y1 or postjunctional receptors that increases blood pressure but has no inhibitory effects on cardiac vagal action. ONE-LETTER SEQUENCE: YPSKPDNPGEDAPAEDLARYYSALRHYINLITRPRY-NH2MOLECULAR FORMULA: C190H286N54O56MOLECULAR WEIGHT:4222.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [128768-54-7], RESEARCH AREA: Neuropeptides
Catalog No. 50-210-9938