Learn More
CPC Scientific H-Tyr-Pro-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Pro-Arg-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PEPY003B
Encompass
ONE-LETTER SEQUENCE: YPIKPEAPGEDASPEELNRYYASLRHYLNLVTRPRY-NH2MOLECULAR FORMULA: C194H294N54O56MOLECULAR WEIGHT:4278.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Gastrointestinal
Catalog No. 50-211-0145