Learn More
Electron Microscopy Sciences Linear PRO 7N Vibrating Tissue Slicer
SDP
Catalog No. 501931225
Encompass_Preferred
The Linear Slicer is the first vibrating blade tissue slicer to feature lateral vibration by linear motor (electromagnet) in the world and raises the basic performance of MS linear slicers. This Linear Slicer Model PRO 7N, which drastically reduces poor vibrations and minimizes the death of nerve cells, will definitely help with research activities. The swing width can be adjusted without changing the vibration frequency. Additionally, the advance speed is adjustable.
Catalog No. 50-193-1225 Supplier Electron Microscopy Sciences Supplier No. 93302
Add to Cart
Add to Cart
Request a Quote
The Linear Slicer is the first vibrating blade tissue slicer to feature lateral vibration by linear motor (electromagnet) in the world and raises the basic performance of MS linear slicers. This Linear Slicer Model PRO 7N, which drastically reduces poor vibrations and minimizes the death of nerve cells, will definitely help with research activities. The swing width can be adjusted without changing the vibration frequency. Additionally, the advance speed is adjustable.