Learn More
Medchemexpress LLC Proadifen (hydrochloride) | 62-68-0 | 99.95% | 1 ML
SDP
Catalog No. 5000411843
Encompass_Preferred
Proadifen hydrochloride, also known as SKF-525A, is a non-competitive Cytochrome P450 inhibitor with an IC50 value of 19 μM. It reduces monoamine oxidase A activity and N,N-dimethyltryptamine metabolism. This compound is promising for research related to metabolism-related diseases, ovarian carcinoma, inflammation, and dopamine neuron-related diseases.
  • Reduces monoamine oxidase A activity
  • Reduces N,N-dimethyltryptamine metabolism in liver microsomes
  • Inhibits N-demethylation and Acridone formation
  • Augments lipopolysaccharide-induced fever
  • Exacerbates prostaglandin E2 levels in rats
  • Promising for research of metabolism-related diseases
  • Promising for research of ovarian carcinoma
  • Promising for research of inflammation
  • Promising for research of dopamine neuron-related diseases
Catalog No. 50-004-11843 Supplier Medchemexpress LLC Supplier No. HYB131110MM/1ML
Proadifen hydrochloride, also known as SKF-525A, is a non-competitive Cytochrome P450 inhibitor with an IC50 value of 19 μM. It reduces monoamine oxidase A activity and N,N-dimethyltryptamine metabolism. This compound is promising for research related to metabolism-related diseases, ovarian carcinoma, inflammation, and dopamine neuron-related diseases.
  • Reduces monoamine oxidase A activity
  • Reduces N,N-dimethyltryptamine metabolism in liver microsomes
  • Inhibits N-demethylation and Acridone formation
  • Augments lipopolysaccharide-induced fever
  • Exacerbates prostaglandin E2 levels in rats
  • Promising for research of metabolism-related diseases
  • Promising for research of ovarian carcinoma
  • Promising for research of inflammation
  • Promising for research of dopamine neuron-related diseases