Learn More
Air Science Usa PCR Workstation, 36" / 900mm Nominal Width, 115V 60Hz
SDP
Catalog No. NC1809368
Encompass_Preferred
Purair PCR Workstation with vertical laminar flow and UV (PCR-36-A), 36" / 900mm nominal width. Polymerase chain reaction (PCR) is a simple and inexpensive technique to make multiple copies of a targeted nucleotide sequence from a DNA sample and to amplify sequences from small samples. This technique is widely used in genetics laboratories that work with DNA and RNA. Because PCR amplification is extremely sensitive to contamination, prevention of contamination requires good laboratory practices to minimize external or cross-contamination during reagent preparation, sample preparation and sample amplification. The Purair PCR laminar flow cabinet employs the Multiplex HEPA filtration technology to create a safe, energy-efficient contaminant-free environment. This PCR workstation is ideally suited for use with non-hazardous contaminants and when flexible access to the instrumentation in the cabinet is required
Catalog No. NC1809368 Supplier Air Science Usa Supplier No. PCR36 A
Purair PCR Workstation with vertical laminar flow and UV (PCR-36-A), 36" / 900mm nominal width. Polymerase chain reaction (PCR) is a simple and inexpensive technique to make multiple copies of a targeted nucleotide sequence from a DNA sample and to amplify sequences from small samples. This technique is widely used in genetics laboratories that work with DNA and RNA. Because PCR amplification is extremely sensitive to contamination, prevention of contamination requires good laboratory practices to minimize external or cross-contamination during reagent preparation, sample preparation and sample amplification. The Purair PCR laminar flow cabinet employs the Multiplex HEPA filtration technology to create a safe, energy-efficient contaminant-free environment. This PCR workstation is ideally suited for use with non-hazardous contaminants and when flexible access to the instrumentation in the cabinet is required