Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Revolutionary Science ProBath 150 (5L) Water Bath
SCI_ED

Catalog No. S01571 Shop All Revolutionary Science Products
Change view
Click to view available options
Capacity (Metric):
5.7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01571 5.7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01571 Supplier Revolutionary Science Supplier No. RSPB150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ProBath 150 delivers performance with a generous 9.5 in. vertical clearance, accommodating tall items without removing the lid.

  • Digital temperature control
  • 5.7 liter, 300 series stainless steel chamber
  • Fits flasks, bottles, and beakers up to 9.5 in.
  • Over and under temperature alarms
  • Simple 3-button operation
  • Slim, space-saving profile
  • Transparent, polycarbonate lid
  • Temperature stability: ±0.4°C
  • Temperature range: Ambient to 90.0°C
  • Great for DNA extraction, CRISPR, heat shock, vaccine and agar thawing, and more
  • 2-year warranty

Specifications

Autoclavable Not Autoclavable
Capacity (English) 1.5 gal.
Color White
Capacity (Metric) 5.7 L
For Use With (Equipment) DNA extraction, CRISPR, heat shock, vaccine and agar thawing, and more
Temperature Range Ambient to 90.0°C
Type Water Bath
For Use With (Application) Warming materials
Includes Lid, Tray
Material Painted Steel
Temperature Stability ±0.4°C
Temperature Uniformity ±0.1°C
Length (English) 15.75 in.
Length (Metric) 40 cm
Width (English) 7 in.
Width (Metric) 17.8 cm
Height (English) 8 in.
Height (Metric) 20.3 cm
Depth (English) 5.875 in.
Depth (Metric) 14.9 cm
Weight (English) 8 lb.
Weight (Metric) 3.63 kg
Electrical Requirements 120 V, 50/60 Hz
Wattage 250 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.