Learn More
Ohaus Probe
Catalog No. 1676277 Shop All OHAUS Products
Change view
Click to view available options
Type:
Temperature Probe
Catalog No. Type
16-762-77 Temperature Probe
1 options
Catalog No. 16-762-77 Supplier Ohaus Supplier No. 30973654
Add to Cart
Add to Cart
Stainless steel temperature probe for digital hotplates and hotplate stirrers.
  • Probe PT100(A) -40 to 400°C
  • 25cm
  • SS 316/304
TRUSTED_SUSTAINABILITY
Specifications
Type Temperature Probe
Warranty 2 Years
Material Stainless Steel