Learn More
Cole-Parmer Digi-Sense Type-K, SS 90guard Surf Probe; 6.5" L, Mini-Connector, Exposed, 3ft FEP Cable
Catalog No. 50200175 Shop All Cole-Parmer Products
Encompass_Preferred
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C)
Catalog No. 50-200-175 Supplier Cole-Parmer Supplier No. 0844741
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C)