Learn More
Bellco Glass, Inc. Probe Adapter, 2.5in.
SDP
Catalog No. 501944951
Encompass_Preferred
316 l stainless steel probe adapter threaded. Not a thermowell. This is a fixed height probe holder for the Omni series headplates. Also requires bottom half of a 1/2" compression fitting A554-33 and A554-42 o-ring. PG 13.5 THREAD X 2.5" EXTENSION. MADE IN THE USA.
Catalog No. 50-194-4951 Supplier Bellco Glass, Inc. Supplier No. 196492005
316 l stainless steel probe adapter threaded. Not a thermowell. This is a fixed height probe holder for the Omni series headplates. Also requires bottom half of a 1/2" compression fitting A554-33 and A554-42 o-ring. PG 13.5 THREAD X 2.5" EXTENSION. MADE IN THE USA.