Learn More
Bellco Glass, Inc. Probe Adapter 3.25"
SDP
Catalog No. 501944952
Encompass_Preferred
316 l stainless steel probe adapter threaded. These thermowells are designed specifically for the Bellco control systems and fit through a 5/16" fitting on the stainless steel headplate. Pg 13.5 thread x 3.25" extension. MADE IN THE USA.
Catalog No. 50-194-4952 Supplier Bellco Glass, Inc. Supplier No. 196492010
316 l stainless steel probe adapter threaded. These thermowells are designed specifically for the Bellco control systems and fit through a 5/16" fitting on the stainless steel headplate. Pg 13.5 thread x 3.25" extension. MADE IN THE USA.